missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD357 (AITR/GITR) Mouse anti-Human, Alexa Fluor 488, Clone: eBioAITR, eBioscience™

Mouse Monoclonal Antibody

$279.84 - $361.68

Specifications

Antigen CD357 (AITR/GITR)
Clone eBioAITR
Host Species Murine
Gene Alias TNFRSF18, activation-inducible TNFR family member
Species Reactivity Human
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5016775 Life Technologies
LS53587541
25 Tests Each for $279.84
5016776 Life Technologies
LS53587542
100 Tests Each for $361.68
Specifications

Description

The monoclonal antibody reacts with human AITR, Activation Inducible TNFR family member, an approximately 25kDa member of the TNFR superfamily. AITR mRNA is detected in lymph node, peripheral blood leukocytes and weakly in spleen. AITR mRNA expression was upregulated within 24 hour on PMA/ionomycin or PHA stimulated PBMC. At the protein level, AITR is expressed by a small population of activated PBMC. AITR associates with TRAF1, TRAF2 and TRAF3 and induces nuclear factor NF-κB activation via TRAF2. Recently TL6 (AITRL) has been reported as the ligand for AITR. Interaction of AITR with AITRL is important for cross-talk between T lymphocytes and endothelial cells.
Description & Specifications

Specifications

CD357 (AITR/GITR)
Murine
Human
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
eBioAITR
TNFRSF18, activation-inducible TNFR family member
Flow Cytometry
Alexa Fluor 488
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
5μL (0.25μg)/test