missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD354 (TREM-1) Rat anti-Mouse, eFluor 660, Clone: TR3MBL1, eBioscience™

Rat Monoclonal Antibody

$157.08 - $394.68

Specifications

Antigen CD354 (TREM-1)
Clone TR3MBL1
Host Species Rat
Species Reactivity Mouse
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501124343 Life Technologies
LS50354180
25μg Each for $157.08
501124344 Life Technologies
LS50354182
100μg Each for $394.68
Specifications

Description

The TR3MBL1 monoclonal antibody recognizes mouse CD354 (also known as TREM-1), a 30kDa glycosylated member of the Ig superfamily. CD354 is expressed on the cell surface of myeloid cells such as neutrophils, monocytes, dendritic cells, some macrophages, platelets, and megakaryocytes. Outside the immune system, CD354 is expressed on hepatocytes, osteoclasts, and microglia. CD354 also exists as a soluble receptor (sTREM-1) that arises from proteolytic cleavage of the membrane-bound receptor by matrix metalloproteases. CD354 plays a critical role in the innate immune response to bacterial and fungal infection by triggering the release of pro-inflammatory chemokines and cytokines. The cell surface receptor associates constitutively with the ITAM-containing adaptor protein DAP12 to activate the MAPK and NFkB signaling pathways. Finally, levels of the soluble receptor are increased in several inflammatory diseases, including sepsis, asthma, and inflammatory bowel disease, making this receptor a valuable diagnostic biomarker.
Description & Specifications

Specifications

CD354 (TREM-1)
Rat
Flow Cytometry
eFluor 660
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.2mg/mL
TR3MBL1
Mouse
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer