Please login to your online account to display your discounted pricing

CD34 Mouse anti-Human, Biotin, Clone: 4H11, eBioscience™

Mouse Monoclonal Antibody

$143.55 - $259.55

Specifications

Antigen CD34
Clone 4H11
Host Species Murine
Gene Alias Mucosialin, Class III epitope
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5011519

 
life technologies
LS13034980
25μg Each for $143.55

5011520

 
life technologies
LS13034982
100μg Each for $259.55
Description & Specifications

Specifications

Antigen CD34
Clone 4H11
Host Species Murine
Gene Alias Mucosialin, Class III epitope
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate Biotin
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

The 4H11 monoclonal antibody reacts with human CD34, also known as mucosialin. CD34 belongs to a protein family which also includes endoglycan and podocalyxin. Members of this family are single pass transmembrane proteins with a heavily glycosylated extracellular and N-terminal mucin domain. CD34 was first identified as an antigen expressed on hematopoietic progenitors, and has since been extensively used as a marker to isolate cells capable of hematopoietic cell engraftment. In spite of this, the function of CD34 remains unresolved. In addition to expression on hematopoietic progenitors, CD34 is expressed on some populations of mesenchymal stem cells, tumor cell lines, and by vascular endothelia in the adult. Epitopes of CD34 have been assigned to three classes (class I, II or III) based on their differential sensitivity to enzymatic cleavage by neuraminidase, chymopapain, or O-glycoprotease. According to this analysis, the 4H11 antibody belongs to class III, indicating that it reacts with a protein epitope.