missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD34 Mouse anti-Canine, PE, Clone: 1H6, eBioscience™

Mouse Monoclonal Antibody

$143.88 - $355.08

Specifications

Antigen CD34
Clone 1H6
Host Species Murine
Species Reactivity Canine
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5010330 Life Technologies
LS12034041
25 Tests Each for $143.88
5010331 Life Technologies
LS12034042
100 Tests Each for $355.08
Specifications

Description

The 1H6 monoclonal antibody reacts with canine CD34, also known as mucosialin. CD34 belongs to a protein family that also includes endoglycan and podocalyxin. These single-pass transmembrane proteins possess heavily glycosylated extracellular and N-terminal mucin domains. Similar to humans, canine CD34 can be detected on hematopoietic progenitor cells in the bone marrow, peripheral blood and hair follicle, as well as on some populations of mesenchymal stem cells, tumor cell lines, and on vascular endothelia in the adult. CD34 can be used as a marker to isolate cells capable of hematopoietic cell engraftment.

The 1H6 antibody has been reported to work for enriching CD34+ cells.
Description & Specifications

Specifications

CD34
Murine
Flow Cytometry
PE
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
5μL (0.25μg)/test
1H6
Canine
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer