Please login to your online account to display your discounted pricing

CD338 (ABCG2) Mouse anti-Human, PE, Clone: 5D3, eBioscience™

Mouse Monoclonal Antibody

$187.05 - $390.05

Specifications

Antigen CD338 (ABCG2)
Clone 5D3
Host Species Murine
Gene Alias Bcrp1, MXR
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501122399

 
life technologies
LS12888841
25 Tests Each for $187.05

501122400

 
life technologies
LS12888842
100 Tests Each for $390.05
Description & Specifications

Specifications

Antigen CD338 (ABCG2)
Clone 5D3
Host Species Murine
Gene Alias Bcrp1, MXR
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL; 5μL (0.25μg)/test

The 5D3 monoclonal antibody reacts with the extracellular portion of the human ABCG2 protein, also known as Bcrp1 and MXR. The ABCG2 gene, a member of the multi-drug resistance (MDR) family, is highly expressed on primitive ‘side-population’ (SP) stem cells, which are defined by the efflux of fluorescent dyes such as Rhodamine 123 and Hoechest 33342. In the bone marrow, about 0.05% of cells display the low fluorescence and are highly enriched for repopulating cells. The SP cells, which express low or undetectable levels of CD34, have been identified in multiple species. In addition, expression of ABCG2 appears to be highly conserved.