missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD337 (NKp30) Mouse anti-Human, PE, Clone: AF29-4D12, eBioscience™

Mouse Monoclonal Antibody

$138.60 - $341.88

Specifications

Antigen CD337 (NKp30)
Clone AF29-4D12
Host Species Murine
Species Reactivity Human, Non-human primate, Rhesus Ma
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501122309 Life Technologies
LS12337941
25 Tests Each for $138.60
501122310 Life Technologies
LS12337942
100 Tests Each for $341.88
Specifications

Description

This AF29-4D12 monoclonal antibody reacts with human CD337, which is also known as NKp30. Expressed on NK cells, CD337 is the predominant member of the natural cytotoxicity receptor family that also includes NKp46 and NKp44. CD337 is involved in activating and expanding resting NK cells following interaction with dendritic cells to eliminate tumor, virus-infected, and immature dendritic cells. Several ligands for this receptor have been reported, including B7-H6.

The AF29-4D12 antibody has been reported to crossreact to rhesus macaque.
Description & Specifications

Specifications

CD337 (NKp30)
Murine
Flow Cytometry
PE
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
5μL (0.125μg)/test
AF29-4D12
Human, Non-human primate, Rhesus Ma
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer