Please login to your online account to display your discounted pricing

CD336 (NKp44) Mouse anti-Human, eFluor 450, Clone: 44.189, eBioscience™

Mouse Monoclonal Antibody

$187.05 - $433.55

Specifications

Antigen CD336 (NKp44)
Clone 44.189
Host Species Mouse
Gene Alias NCR2
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501124157

 
life technologies
LS48336941
25 tests Each for $187.05

501124158

 
life technologies
LS48336942
100 tests Each for $433.55
Description & Specifications

Specifications

Antigen CD336 (NKp44)
Clone 44.189
Host Species Mouse
Gene Alias NCR2
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate eFluor 450
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (1.0μg)/test

The monoclonal antibody 44.189 reacts with human CD336, which is also known as NKp44 or NCR2. CD336 is a 44kDa transmembrane glycoprotein belonging to the natural cytotoxicity receptor (NCR) family and contains an ITIM domain located in the cytoplasmic tail. Expression of CD336 and NK cell activation by the receptor requires association with DAP12, an ITAM-containing transmembrane accessory protein. Once thought to be expressed exclusively on IL-2-activated NK cells and a subset of gamma delta T lymphocytes, studies have shown expression of CD336 on IL-3-stimulated plasmacytoid dendritic cells, alpha beta T cells in the small intestinal epithelium, and T cells in cord blood. CD336 has been shown to be involved in the innate response of activated NK cells against viral infection and tumor cell proliferation.

The 44.189 antibody has been reported to facilitate CD336 crosslinking.