missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD336 (NKp44) Mouse anti-Human, APC, Clone: 44.189, eBioscience™

Mouse Monoclonal Antibody

$157.08 - $368.28

Specifications

Antigen CD336 (NKp44)
Clone 44.189
Host Species Murine
Gene Alias NCR2
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501123125 Life Technologies
LS17336941
25 Tests Each for $157.08
501123126 Life Technologies
LS17336942
100 Tests Each for $368.28
Specifications

Description

The monoclonal antibody 44.189 reacts with human CD336, which is also known as NKp44 or NCR2. CD336 is a 44kDa transmembrane glycoprotein belonging to the natural cytotoxicity receptor (NCR) family and contains an ITIM domain located in the cytoplasmic tail. Expression of CD336 and NK cell activation by the receptor requires association with DAP12, an ITAM-containing transmembrane accessory protein. Once thought to be expressed exclusively on IL-2-activated NK cells and a subset of gamma delta T lymphocytes, studies have shown expression of CD336 on IL-3-stimulated plasmacytoid dendritic cells, alpha beta T cells in the small intestinal epithelium, and T cells in cord blood. CD336 has been shown to be involved in the innate response of activated NK cells against viral infection and tumor cell proliferation.

The 44.189 antibody has been reported to facilitate CD336 crosslinking.
Description & Specifications

Specifications

CD336 (NKp44)
Murine
Flow Cytometry
Allophycocyanin
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
5μL (0.25ugμg)/test
44.189
NCR2
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer