missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD335 (NKp46), PE-eFluor 610, clone: 29A1.4, eBioscience™

Rat Monoclonal Antibody

Manufacturer:  Life Technologies LS61335180

 View more versions of this product

Catalog No. 501124806



Description

Description

Cytotoxicity-activating receptor that may contribute to the increased efficiency of activated natural killer (NK) cells to mediate tumor cell lysis.

PE-eFluor® 610 can be excited with laser lines from 488-561 nm and emits at 607 nm. We recommend using a 610/20 band pass filter (equivalent to PE-Texas Red®). Please make sure that your instrument is capable of detecting this fluorochome. Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 µL of cell sample + 100 µL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically. Excitation: 488-561 nm; Emission: 607 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered. Cytotoxicity-activating receptor that may contribute to the increased efficiency of activated natural killer (NK) cells to mediate tumor cell lysis.
Specifications

Specifications

CD335 (NKp46)
Monoclonal
0.2 mg/mL
PBS with 0.09% sodium azide; pH 7.2
Q8C567
CD335, LY94, NK-p46, NKP46, NK cell-activating receptor, lymphocyte antigen 94 homolog activating NK-receptor, NK-p46, natural killer cell p46-related protein
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Mouse
Flow Cytometry
29A1.4
PE-eFluor® 610
Ncr1
Liquid
Ncr1
IgG2a, kappa
25 μg
4° C, store in dark, DO NOT FREEZE!
Primary
17086
Documents

For Research Use Only.