missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD335 (NKp46) Rat anti-Mouse, APC-eFluor® 780, Clone: 29A1.4, eBioscience™

Rat Monoclonal Antibody

Manufacturer:  Life Technologies LS47335180

 View more versions of this product

Catalog No. 501129221



Description

Description

Description: The monoclonal antibody 29A1.4 recognizes mouse NKp46, also known as CD335. CD335, a member of the natural cytotoxicity receptor (NCR) family, is a glycoprotein with 2 Ig-like domains and a short cytoplasmic tail. Expression of CD335 is uniquely found on NK cells (including immature NK cells, defined as DX5- CD3-, and thereby allowing discrimination between NKT cells and NK cells (NKp46+, CD3-). Furthermore, unlike many of the NK markers which also stain NKT cells, staining with 29A1.4 is not strain specific. Staining has been shown on C57Bl/6, SJL, CBA/CA and BALB/C strains. NKp46 has been shown to play a role in NK cell-mediated lysis of several tumor cells and pathogen-infected cell lines. The 29A1.4 monoclonal antibody has been shown to activate NK cells in vitro. The 29A1.4 monoclonal antibody does not deplete NK cells in vivo. Applications Reported: This 29A1.4 antibody has been reported for use in flow cytometric analysis. Applications Tested: This 29A1.4 antibody has been tested by flow cytometric analysis of mouse spleen cells. This can be used at less than or equal to 1 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

APC-eFluor® 780 emits at 780 nm and is excited with the Red laser (633 nm). Please make sure that your instrument is capable of detecting this fluorochome. Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 µL of cell sample + 100 µL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically. Excitation: 633-647 nm; Emission: 780 nm; Laser: Red Laser. Filtration: 0.2 μm post-manufacturing filtered. Cytotoxicity-activating receptor that may contribute to the increased efficiency of activated natural killer (NK) cells to mediate tumor cell lysis.
Specifications

Specifications

CD335 (NKp46)
Monoclonal
0.2 mg/mL
PBS with 0.09% sodium azide; pH 7.2
Q8C567
CD335, LY94, NK-p46, NKP46, NK cell-activating receptor, lymphocyte antigen 94 homolog activating NK-receptor, NK-p46, natural killer cell p46-related protein
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Mouse
Flow Cytometry
29A1.4
APC-eFluor® 780
Ncr1
Liquid
Ncr1
IgG2a, kappa
25 μg
4° C, store in dark, DO NOT FREEZE!
Primary
17086
Documents

For Research Use Only.