Please login to your online account to display your discounted pricing

CD335 (NKp46) Mouse anti-Human, PerCP-eFluor 710, Clone: 9E2, eBioscience™

Mouse Monoclonal Antibody

$201.55 - $491.55

Specifications

Antigen CD335 (NKp46)
Clone 9.00E+02
Host Species Mouse
Gene Alias NCR1, Ly-94
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501129337

 
life technologies
LS46335942
100 tests Each for $491.55

501130802

 
life technologies
LS46335941
25 tests Each for $201.55
Description & Specifications

Specifications

Antigen CD335 (NKp46)
Clone 9.00E+02
Host Species Mouse
Gene Alias NCR1, Ly-94
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PerCP-eFluor 710
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.125μg)/test

This 9E2 monoclonal antibody reacts with human CD335, which is also known as NKp46. Expressed on NK cells, CD335 is a member of the natural cytotoxicity receptor family that also includes NKp30 and NKp44. This type I transmembrane glycoprotein is involved in activating and expanding resting NK cells following interaction with dendritic cells to eliminate tumor and virus-infected (e.g., influenza) cells. CD335 also interacts with pancreatic beta cells during the development of type I diabetes.

The 9E2 antibody has been reported to induce cytotoxicity.