missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD335 (NKp46) Mouse anti-Human, eFluor 450, Clone: 9E2, eBioscience™

Mouse Monoclonal Antibody

$145.20 - $361.68

Specifications

Antigen CD335 (NKp46)
Clone 9.00E+02
Host Species Murine
Gene Alias NCR1, Ly-94
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501124155 Life Technologies
LS48335941
25 Tests Each for $145.20
501124156 Life Technologies
LS48335942
100 Tests Each for $361.68
Specifications

Description

This 9E2 monoclonal antibody reacts with human CD335, which is also known as NKp46. Expressed on NK cells, CD335 is a member of the natural cytotoxicity receptor family that also includes NKp30 and NKp44. This type I transmembrane glycoprotein is involved in activating and expanding resting NK cells following interaction with dendritic cells to eliminate tumor and virus-infected (e.g., influenza) cells. CD335 also interacts with pancreatic beta cells during the development of type I diabetes.

The 9E2 antibody has been reported to induce cytotoxicity.
Description & Specifications

Specifications

CD335 (NKp46)
Murine
Flow Cytometry
eFluor 450
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
5μL (0.125μg)/test
9.00E+02
NCR1, Ly-94
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer