Please login to your online account to display your discounted pricing

CD335 (NKp46) Mouse anti-Human, Clone: 9E2, Functional Grade, eBioscience™

Mouse Monoclonal Antibody, Functional Grade

$259.55 - $578.55

Specifications

Antigen CD335 (NKp46)
Clone 9.00E+02
Host Species Mouse
Gene Alias NCR1, Ly-94
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501122954

 
life technologies
LS16335982
100μg Each for $259.55

501122955

 
life technologies
LS16335985
500μg Each for $578.55
Description & Specifications

Specifications

Antigen CD335 (NKp46)
Clone 9.00E+02
Host Species Mouse
Gene Alias NCR1, Ly-94
Species Reactivity Human
Applications Flow Cytometry
Applications Functional Assay
Regulatory Status RUO
Conjugate Unlabeled
Format Purified
Storage Requirements Store at 2-8°C.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, no sodium azide
Concentration 1mg/mL

This 9E2 monoclonal antibody reacts with human CD335, which is also known as NKp46. Expressed on NK cells, CD335 is a member of the natural cytotoxicity receptor family that also includes NKp30 and NKp44. This type I transmembrane glycoprotein is involved in activating and expanding resting NK cells following interaction with dendritic cells to eliminate tumor and virus-infected (e.g., influenza) cells. CD335 also interacts with pancreatic beta cells during the development of type I diabetes.

The 9E2 antibody has been reported to induce cytotoxicity.