Learn More
CD33 Monoclonal Antibody (9A11-CD33), PE-Cyanine7, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
Supplier: Invitrogen 25033182
Description
Description: This monoclonal antibody 9A11-CD33 reacts with mouse CD33, also known as Siglec3 and gp67. CD33 belongs to the sialic acid-binding immunoglobulin lectins (Siglec) family that binds to sialic acid-containing glycans on cell surface glycoproteins and glycolipids on mammalian cells. Mouse CD33 binds to short O-linked glycans of certain mucins and sialyl-Tn epitopes. Mouse CD33 is expressed by mature granulocytes, immature myeloid cells, and microglia. Unlike human CD33, mouse CD33 lacks a cytoplasmic ITIM domain and exists as two splice variants that have different cytoplasmic tails. In microglia, CD33 inhibits the phagocytosis of toxic amyloid beta. Applications Reported: This 9A11-CD33 antibody has been reported for use in flow cytometric analysis. Applications Tested: This 9A11-CD33 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (cat.
CD33 Monoclonal antibody specifically detects CD33 in Mouse samples. It is validated for Flow Cytometry.Specifications
CD33 | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
Q63994 | |
CD33 | |
Affinity chromatography | |
RUO | |
12489 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
9A11-CD33 | |
PE-Cyanine7 | |
CD33 | |
CD33; CD33 antigen; CD33 antigen (gp67); CD33 molecule; CD33 molecule transcript; FLJ00391; gp67; Myeloid cell surface antigen CD33; p67; sialic acid binding Ig-like lectin 3; sialic acid-binding Ig-like lectin 3; SIGLEC3; Siglec-3 | |
Rat | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG1 κ |
For Research Use Only.