Please login to your online account to display your discounted pricing

CD33 Mouse anti-Human, PE-Cy5, Clone: HIM3-4, eBioscience™

Mouse Monoclonal Antibody

$172.55 - $375.55

Specifications

Antigen CD33
Clone HIM3-4
Host Species Mouse
Gene Alias Sialic Acid-Binding Immunoglobulin-Like Lectin 3, SIGLEC3
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5014041

 
affymetrix
15033941
25 tests Each for $172.55

5014042

 
affymetrix
15033942
100 tests Each for $375.55
Description & Specifications

Specifications

Antigen CD33
Clone HIM3-4
Host Species Mouse
Gene Alias Sialic Acid-Binding Immunoglobulin-Like Lectin 3, SIGLEC3
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE-Cy5
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (1μg)/test

The HIM3-4 monoclonal antibody reacts with human CD33, a 67kDa member of the sialoadhesion family. CD33 is expressed by myelomonocytic precursors, monocytes, mast cells, and granulocytes. Hematopoietic stem cells and lymphocytes do not express this antigen. CD33 plays a role in sialic-acid dependent cell adhesion.