missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD33 Mouse anti-Human, PE, Clone: WM-53 (WM53), eBioscience™

Mouse Monoclonal Antibody

$130.68 - $289.08

Specifications

Antigen CD33
Clone WM-53 (WM53)
Host Species Murine
Gene Alias Sialic Acid-Binding Immunoglobulin-Like Lectin 3, SIGLEC3
Species Reactivity Human
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5010326 Life Technologies
LS12033841
25 Tests Each for $130.68
5010327 Life Technologies
LS12033842
100 Tests Each for $289.08
Specifications

Description

The WM-53 monoclonal antibody reacts with human CD33, also known as GP67 and P67, a 67kDa type I transmembrane glycoprotein that is a member of the Siglec (sialic acid-binding Ig superfamily lectin) family. It is highly specific to the hematopoietic compartment and is expressed on monocytes, activated T cells, granulocytes, myeloid progenitors, and mast cells.
Description & Specifications

Specifications

CD33
Murine
Human
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
WM-53 (WM53)
Sialic Acid-Binding Immunoglobulin-Like Lectin 3, SIGLEC3
Flow Cytometry
PE
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
5μL (0.25μg)/test