missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD33 Mouse anti-Human, PE, Clone: HIM3-4, eBioscience™

Mouse Monoclonal Antibody

$145.00 - $324.80

Specifications

Antigen CD33
Clone HIM3-4
Host Species Murine
Gene Alias Sialic Acid-Binding Immunoglobulin-Like Lectin 3, SIGLEC3
Species Reactivity Human
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5010328 Life Technologies
LS12033941
25 Tests Each for $145.00
5010329 Life Technologies
LS12033942
100 Tests Each for $324.80
Specifications

Description

The HIM3-4 monoclonal antibody reacts with human CD33, a 67kDa member of the sialoadhesion family. CD33 is expressed by myelomonocytic precursors, monocytes, mast cells, and granulocytes. Hematopoietic stem cells and lymphocytes do not express this antigen. CD33 plays a role in sialic-acid dependent cell adhesion.
Description & Specifications

Specifications

CD33
Murine
Human
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
HIM3-4
Sialic Acid-Binding Immunoglobulin-Like Lectin 3, SIGLEC3
Flow Cytometry
PE
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
5μL (0.5μg)/test