Please login to your online account to display your discounted pricing

CD33 Mouse anti-Human, FITC, Clone: WM-53 (WM53), eBioscience™

Mouse Monoclonal Antibody

$114.55 - $266.80

Specifications

Antigen CD33
Clone WM-53 (WM53)
Host Species Mouse
Gene Alias Sialic Acid-Binding Immunoglobulin-Like Lectin 3, SIGLEC3
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501122060

 
life technologies
LS11033841
25 tests Each for $114.55

501122061

 
life technologies
LS11033842
100 tests Each for $266.80
Description & Specifications

Specifications

Antigen CD33
Clone WM-53 (WM53)
Host Species Mouse
Gene Alias Sialic Acid-Binding Immunoglobulin-Like Lectin 3, SIGLEC3
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate FITC
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.5μg)/test

The WM-53 monoclonal antibody reacts with human CD33, also known as GP67 and P67, a 67kDa type I transmembrane glycoprotein that is a member of the Siglec (sialic acid-binding Ig superfamily lectin) family. It is highly specific to the hematopoietic compartment and is expressed on monocytes, activated T cells, granulocytes, myeloid progenitors, and mast cells.