Please login to your online account to display your discounted pricing

CD33 Mouse anti-Human, eFluor 450, Clone: P67.6, eBioscience™

Mouse Monoclonal Antibody

$201.55 - $448.05

Specifications

Antigen CD33
Clone P67.6
Host Species Mouse
Gene Alias Sialic Acid-Binding Immunoglobulin-Like Lectin 3, SIGLEC3
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5016284

 
life technologies
LS48033741
25 tests Each for $201.55

501128803

 
life technologies
LS48033742
100 tests Each for $448.05
Description & Specifications

Specifications

Antigen CD33
Clone P67.6
Host Species Mouse
Gene Alias Sialic Acid-Binding Immunoglobulin-Like Lectin 3, SIGLEC3
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate eFluor 450
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.25μg)/test

The P67.6 monoclonal antibody reacts with human CD33 (also known as GP67 and P67), a 67kDa type I transmembrane glycoprotein that is a member of the Siglec (sialic acid-binding Ig superfamily lectin) family. It is highly specific to the hematopoietic compartment and is expressed on monocytes, activated T cells, granulocytes, myeloid progenitors, and mast cells.