Please login to your online account to display your discounted pricing

CD33 Mouse anti-Human, Clone: WM-53 (WM53), eBioscience™

Mouse Monoclonal Antibody

$71.05 - $158.05

Specifications

Antigen CD33
Clone WM-53 (WM53)
Host Species Murine
Gene Alias Sialic Acid-Binding Immunoglobulin-Like Lectin 3, SIGLEC3
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501122642

 
life technologies
LS14033882
100μg Each for $158.05

5012527

 
life technologies
LS14033880
25μg Each for $71.05
Description & Specifications

Specifications

Antigen CD33
Clone WM-53 (WM53)
Host Species Murine
Gene Alias Sialic Acid-Binding Immunoglobulin-Like Lectin 3, SIGLEC3
Species Reactivity Human
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunoprecipitation
Regulatory Status RUO
Conjugate Unlabeled
Format Purified
Storage Requirements Store at 2-8°C.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

The WM-53 monoclonal antibody reacts with human CD33, also known as GP67 and P67, a 67kDa type I transmembrane glycoprotein that is a member of the Siglec (sialic acid-binding Ig superfamily lectin) family. It is highly specific to the hematopoietic compartment and is expressed on monocytes, activated T cells, granulocytes, myeloid progenitors, and mast cells.