missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD326 (EpCAM) Rat anti-Mouse, Clone: G8.8, eBioscience™

Rat Monoclonal Antibody

$91.08 - $355.08

Specifications

Antigen CD326 (EpCAM)
Clone G8.8
Host Species Rat
Gene Alias epithelial cell adhesion molecule
Species Reactivity Mouse
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5013158 Life Technologies
LS14579181
50μg Each for $91.08
5013160 Life Technologies
LS14579185
500μg Each for $355.08
Specifications

Description

The G8.8 monoclonal antibody reacts with the 40kDa mouse EpCAM (epithelial cellular adhesion molecule), also known as EGP40 (epithelial glycoprotein 40), 17-1A antigen, TACSTD1 (tumor-associated calcium signal transducer 1), and CD326. The immunogen used to generate the G8.8 antibody was the TE-71 thymic epithelial cell line. CD326 is expressed on the majority of epithelial cells, and is considered a pan-carcinoma antigen. CD326 mediates calcium-independent, homophilic, cell-cell adhesion and may function as a growth factor receptor. The antigen is being used as a target for immunotherapy treatment of human carcinomas. CD326 binds LAIR-1 (CD305) and LAIR-2 (CD306) to inhibit cellular activation and inflammation. This epithelial glycoprotein is now recognized as having an important role in tumor biology.
Description & Specifications

Specifications

CD326 (EpCAM)
Rat
Mouse
RUO
Purified
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
G8.8
epithelial cell adhesion molecule
Flow Cytometry,Immunohistochemistry (Frozen),Immunoprecipitation
Unlabeled
Store at 2-8°C.
Monoclonal
0.5mg/mL