Please login to your online account to display your discounted pricing

CD326 (EpCAM) Mouse anti-Human, PE-Cy7, Clone: 1B7, eBioscience™

Mouse Monoclonal Antibody

$200.10 - $501.70

Specifications

Antigen CD326 (EpCAM)
Clone 1B7
Host Species Mouse
Gene Alias Epithelial cell adhesion molecule, KSA, TROP1
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501123450

 
life technologies
LS25932641
25 tests Each for $200.10

501128652

 
life technologies
LS25932642
100 tests Each for $501.70
Description & Specifications

Specifications

Antigen CD326 (EpCAM)
Clone 1B7
Host Species Mouse
Gene Alias Epithelial cell adhesion molecule, KSA, TROP1
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE-Cy7
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.06μg)/test

EpCAM (Epithelial cell adhesion molecule, CD326, KSA, TROP1) is a 40 kD cell-surface adhesion molecule participating in homophilic, calcium-independent cell-cell interactions. EpCAM is a type-I transmembrane protein, and is expressed primarily on the basolateral surface of most epithelia. Although normal epithelia express low levels of EpCAM, increased expression has been correlated with increased proliferation and progression to a mesenchymal phenotype. EpCAM has also been used as a diagnostic marker on circulating metastatic carcinoma cells, while cancer cells of non-epithelial origin do not express EpCAM.