Please login to your online account to display your discounted pricing

CD326 (EpCAM) Mouse anti-Human, Biotin, Clone: 1B7, eBioscience™

Mouse Monoclonal Antibody

$159.50 - $339.30

Specifications

Antigen CD326 (EpCAM)
Clone 1B7
Host Species Mouse
Gene Alias Epithelial cell adhesion molecule, KSA, TROP1
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5012249

 
life technologies
LS13932680
25μg Each for $159.50

501128712

 
life technologies
LS13932682
100μg Each for $339.30
Description & Specifications

Specifications

Antigen CD326 (EpCAM)
Clone 1B7
Host Species Mouse
Gene Alias Epithelial cell adhesion molecule, KSA, TROP1
Species Reactivity Human
Applications Flow Cytometry
Applications Immunohistochemistry
Regulatory Status RUO
Conjugate Biotin
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

EpCAM (Epithelial cell adhesion molecule, CD326, KSA, TROP1) is a 40 kD cell-surface adhesion molecule participating in homophilic, calcium-independent cell-cell interactions. EpCAM is a type-I transmembrane protein, and is expressed primarily on the basolateral surface of most epithelia. Although normal epithelia express low levels of EpCAM, increased expression has been correlated with increased proliferation and progression to a mesenchymal phenotype. EpCAM has also been used as a diagnostic marker on circulating metastatic carcinoma cells, while cancer cells of non-epithelial origin do not express EpCAM.