Please login to your online account to display your discounted pricing

CD325 (N-cadherin) Mouse anti-Human, PE, Clone: 8C11, eBioscience™

Mouse Monoclonal Antibody

$158.05 - $361.05

Specifications

Antigen CD325 (N-cadherin)
Clone 8C11
Host Species Murine
Gene Alias neural cadherin
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501122307

 
life technologies
LS12325942
100 Tests Each for $361.05

501122306

 
life technologies
LS12325941
25 Tests Each for $158.05
Description & Specifications

Specifications

Antigen CD325 (N-cadherin)
Clone 8C11
Host Species Murine
Gene Alias neural cadherin
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.25μg)/test

The 8C11 monoclonal antibody reacts with human CD325, also known as N-Cadherin. CD325 is a 130kDa member of the Cadherin superfamily, and consists of five extracellular repeats, a transmembrane domain and a cytoplasmic domain. CD325 deficient mice die at day 10 of gestation and embryos display major heart defects and malformed neural tubes and somites. Consistent with this, CD325 has been implicated in several aspects of cardiac development including the precardiac mesoderm, establishment of left-right symmetry and cardiac looping morphogenesis. Furthermore, CD325 is normally involved in inducing cell cycle arrest and its expression is frequently dysregulated in cancer cells.