missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD325 (N-cadherin) Mouse anti-Human, Clone: 8C11, eBioscience™

Mouse Monoclonal Antibody

$143.55 - $157.08

Specifications

Antigen CD325 (N-cadherin)
Clone 8C11
Host Species Murine
Gene Alias neural cadherin
Applications Flow Cytometry,Immunohistochemistry,Immunoprecipitation,Western Blot
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501122673 Life Technologies
LS14325980
25μg Each for $143.55
5013018 Life Technologies
LS14325982
100μg Each for $157.08
Specifications

Description

The 8C11 monoclonal antibody reacts with human CD325, also known as N-Cadherin. CD325 is a 130kDa member of the Cadherin superfamily, and consists of five extracellular repeats, a transmembrane domain and a cytoplasmic domain. CD325 deficient mice die at day 10 of gestation and embryos display major heart defects and malformed neural tubes and somites. Consistent with this, CD325 has been implicated in several aspects of cardiac development including the precardiac mesoderm, establishment of left-right symmetry and cardiac looping morphogenesis. Furthermore, CD325 is normally involved in inducing cell cycle arrest and its expression is frequently dysregulated in cancer cells.
Description & Specifications

Specifications

CD325 (N-cadherin)
Murine
Flow Cytometry,Immunohistochemistry,Immunoprecipitation,Western Blot
Unlabeled
Store at 2-8°C.
Monoclonal
0.5mg/mL
8C11
neural cadherin
RUO
Purified
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer