missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD323 Mouse anti-Human, eFluor 570, Clone: SHM33, eBioscience™

Mouse Monoclonal Antibody

$159.72 - $402.60

Specifications

Antigen CD323
Clone SHM33
Host Species Murine
Gene Alias JAM-C
Species Reactivity Human
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501123599 Life Technologies
LS41323980
25μg Each for $159.72
501123600 Life Technologies
LS41323982
100μg Each for $402.60
Specifications

Description

The SHM33 monoclonal antibody reacts with human CD323, which is also known as Junctional Adhesion Molecule (JAM) 3. A member of the Ig superfamily of transmembrane proteins, CD323 is expressed on endothelial and epithelial cells, smooth muscle cells, peripheral nerves, platelets, and some B cells. In addition, this receptor is expressed on VEGF and histamine-stimulated endothelial cells, as well as on activated T cells. CD323 is involved in neutrophil transmigration during inflammation, angiogenesis, cell polarity, and nerve conduction. CD323 interacts homo- and heterotypically with beta2 and beta3 integrins, JAM2, and the viral receptor CAR.
Description & Specifications

Specifications

CD323
Murine
Human
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
SHM33
JAM-C
Immunocytochemistry,Immunohistochemistry,Microscopy
eFluor 570
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.2mg/mL