missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD321 (F11R) Mouse anti-Human, Clone: WK9, eBioscience™

Mouse Monoclonal Antibody

$117.48 - $262.68

Specifications

Antigen CD321 (F11R)
Clone WK9
Host Species Murine
Gene Alias F11 R, JAM-1, JAM-A
Applications Flow Cytometry,Immunoprecipitation,Western Blot
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5013814 Life Technologies
LS14932182
100μg Each for $262.68
5013813 Life Technologies
LS14932180
25μg Each for $117.48
Specifications

Description

CD321 (F11R, JAM-1, JAM-A) is a cell-surface adhesion molecule belonging to the Ig superfamily of receptors. CD321 is a 32-41 kD glycoprotein, possessing two extracellular Ig-like domains, a transmembrane region, and a short cytosolic tail. The protein assembles at the membrane as a homodimer, and can associate laterally with other membrane proteins including integrins. CD321 is expressed on a variety of cell types, including leukocytes, platelets, erythrocytes, epithelial and endothelial cells, and plays a role in mediating cell-cell contact. CD321 can be found in close proximity to several components of epithelial tight junctions, including ZO-1 and occludin. CD321 is also involved in reovirus entry into cells.
Description & Specifications

Specifications

CD321 (F11R)
Murine
Flow Cytometry,Immunoprecipitation,Western Blot
Unlabeled
Store at 2-8°C.
Monoclonal
0.5mg/mL
WK9
F11 R, JAM-1, JAM-A
RUO
Purified
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer