missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD32 Mouse anti-Human, FITC, Clone: 6C4 (CD32), eBioscience™

Mouse Monoclonal Antibody

$104.28 - $283.80

Specifications

Antigen CD32
Clone 6C4 (CD32)
Host Species Murine
Gene Alias Low Affinity IgG Receptor 2a, FcGR2a, IGFR2
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501122059 Life Technologies
LS11032941
25 Tests Each for $104.28
501128653 Life Technologies
LS11032942
100 Tests Each for $283.80
Specifications

Description

This 6C4 monoclonal antibody reacts with human CD32 (also known as Fc gammaRII). This antibody recognizes two isoforms of the receptor, Fc gammaRIIA and Fc gammaRIIB. These 40kDa polymorphic transmembrane glycoproteins are expressed on B cells, granulocytes, monocytes, macrophages, and platelets. Moreover, these receptors are detected on natural killer cells. CD32 enables interaction between Fc gammaRII-expressing cells and opsonized antigen or IgG-containing immune complexes. Both receptors exhibit low affinity towards IgG and play a role in inflammation and autoimmune disease. This clone has been reported to inhibit Ig binding in a rosette blocking assay.
Description & Specifications

Specifications

CD32
Murine
Flow Cytometry
FITC
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
5μL (0.25μg)/test
6C4 (CD32)
Low Affinity IgG Receptor 2a, FcGR2a, IGFR2
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer