missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD317 (BST2, PDCA-1), PE-eFluor 610, clone: eBio927, eBioscience™

Rat Monoclonal Antibody

Manufacturer:  Life Technologies LS61317280

 View more versions of this product

Catalog No. 501124804



Description

Description

BST2, also known as CD317, HM1.24 antigen, DAMP-2 and recently designated tetherin, is an integral membrane protein with a novel topology. The specific function of the BST2, encoded by the bone marrow stromal cell antigen 2 gene, is undetermined; however, it may play a role in pre-B-cell growth, rheumatoid arthritis, and in antiretroviral defense, that blocks release of retrovirus from the cell surface. BST2 is highly expressed during B-cell development, from pro-B precursors to plasma cells, T-cells, monocytes, NK cells and DCs (at protein level).

Description: The eBio927 monoclonal antibody reacts with PDCA-1 (BST2, CD317), a specific marker of plasmacytoid dendritic cells (pDC), also known as type I IFN-producing cells (IPC) in the naive mouse. Mouse IPCs are typically CD11c+, CD11b-, B220+, Ly-6C+, and CD62L+. PDCA-1 is predominantly expressed by IPCs in the naive mouse which represents a very minor population (<0.5%) of splenocytes. It is upregulated on numerous cell types following stimulation which triggers an IFN response. PDCA-1 cycles between cell surface and intracellular compartments and may function to regulate trafficking of secreted cytokines. PDCA-1 (BST2) is the protein recognized by antibody 120G8. The eBio927 monoclonal antibody has also been shown to have functional activity. The epitope recognized by eBio927 is distinct from eBio129c; thus, the antibodies can be used to costain, purify and identify pDCs.Applications Reported: This eBio927 antibody has been reported for use in flow cytometric analysis.Applications Tested: This eBio927 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.PE-eFluor™ 610 can be excited with laser lines from 488-561 nm and emits at 607 nm. We recommend using a 610/20 band pass filter (equivalent to PE-Texas Red™). Please make sure that your instrument is capable of detecting this fluorochome.Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light.Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 μL of cell sample + 100 μL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically.Excitation: 488-561 nm; Emission: 607 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD317 (BST2, PDCA-1)
Monoclonal
0.2mg/mL
PBS with 0.09% sodium azide; pH 7.2
Q8R2Q8
Bone marrow stromal cell antigen 2, Tetherin, PDCA-1, 2310015I10Rik, Bst-2, C87040, DAMP-1, GREG, DAMP-1 protein homolog, HM1.24 antigen, bone marrow stromal antigen 2
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Murine
Flow Cytometry
eBio927
PE-eFluor 610
Bst2
Liquid
Bst2
IgG2b, kappa, kappa
25μg
Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage, handling and experimental procedures.
Primary
69550
Documents

For Research Use Only.