missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD317 (BST2/PDCA-1) Rat anti-Mouse, eFluor 450, Clone: eBio927, eBioscience™

Rat Monoclonal Antibody

$183.48 - $394.68

Specifications

Antigen CD317 (BST2/PDCA-1)
Clone eBio927
Host Species Rat
Gene Alias plasmacytoid DC Ag-1
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5016340 Life Technologies
LS48317280
25μg Each for $183.48
5016341 Life Technologies
LS48317282
100μg Each for $394.68
Specifications

Description

The eBio927 monoclonal antibody reacts with PDCA-1 (BST2, CD317), a specific marker of plasmacytoid dendritic cells (pDC), also known as type I IFN-producing cells (IPC) in the naïve mouse. Mouse IPCs are typically CD11c+, CD11b-, B220+, Ly-6C+, and CD62L+. PDCA-1 is predominantly expressed by IPCs in the naïve mouse which represents a very minor population (<0.5%) of splenocytes. It is upregulated on numerous cell types following stimulation which triggers an IFN response. PDCA-1 cycles between cell surface and intracellular compartments and may function to regulate trafficking of secreted cytokines. PDCA-1 (BST2) is the protein recognized by antibody 120G8.

The eBio927 monoclonal antibody has also been shown to have functional activity. The epitope recognized by eBio927 is distinct from eBio129c; thus, the antibodies can be used to costain, purify and identify pDCs.
Description & Specifications

Specifications

CD317 (BST2/PDCA-1)
Rat
Flow Cytometry
eFluor 450
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.2mg/mL
eBio927
plasmacytoid DC Ag-1
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer