missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD317 (BST2/PDCA-1) Mouse anti-Human, Clone: 26F8, Functional Grade, eBioscience™

Mouse Monoclonal Antibody, Functional Grade

$157.08 - $341.88

Specifications

Antigen CD317 (BST2/PDCA-1)
Clone 26F8
Host Species Murine
Species Reactivity Human
Applications Flow Cytometry,Functional Assay
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5014518 Life Technologies
LS16317980
25μg Each for $157.08
5014519 Life Technologies
LS16317982
100μg Each for $341.88
Specifications

Description

This 26F8 monoclonal antibody reacts with human CD317 (also known as BST2 and tetherin). CD317 is a 30-36kDa type II transmembrane protein expressed on B cells and bone marrow stromal cells. Although reports have indicated that CD317 mRNA is detectable in activated T cells, protein expression in primary T cells and macrophages is undetectable. Moreover, certain T cell lines, such as Jurkat, do not express detectable levels of CD317 protein. CD317 has been associated with pre-B cell growth and the terminal differentiation of plasma B cells. More recently, this molecule has been reported to prevent HIV-1 virion release from the surface of infected cells, leading to reuptake and degradation of the virus. This activity is inhibited by the HIV-1 accessory protein Vpu. CD317 has been identified as the ligand for the ILT7 receptor.
Description & Specifications

Specifications

CD317 (BST2/PDCA-1)
Murine
Flow Cytometry,Functional Assay
Unlabeled
Store at 2-8°C.
Monoclonal
1mg/mL
26F8
Human
RUO
Purified
Primary
aqueous buffer, no sodium azide