Please login to your online account to display your discounted pricing

CD317 (BST2/PDCA-1) Mouse anti-Human, Alexa Fluor 488, Clone: 26F8, eBioscience™

Mouse Monoclonal Antibody

$462.55 - $462.55

Specifications

Antigen CD317 (BST2/PDCA-1)
Clone 26F8
Host Species Murine
Species Reactivity Human
Applications Flow Cytometry
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5016738

 
life technologies
LS53317942
100 Tests Each for $462.55
Description & Specifications

Specifications

Antigen CD317 (BST2/PDCA-1)
Clone 26F8
Host Species Murine
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate Alexa Fluor 488
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.25μg)/test

This 26F8 monoclonal antibody reacts with human CD317 (also known as BST2 and tetherin). CD317 is a 30-36kDa type II transmembrane protein expressed on B cells and bone marrow stromal cells. Although reports have indicated that CD317 mRNA is detectable in activated T cells, protein expression in freshly isolated primary T cells and macrophages is undetectable. Moreover, certain T cell lines, such as Jurkat, do not express detectable levels of CD317 protein. However, CD317 expression can be induced in many cell types (e.g., T and B cells, 293T, and HeLa) by IFNα treatment. CD317 has been associated with pre-B cell growth and the terminal differentiation of plasma B cells. More recently, this molecule has been reported to prevent HIV-1 virion release from the surface of infected cells, leading to reuptake and degradation of the virus. This activity is inhibited by the HIV-1 accessory protein Vpu. CD317 has been identified as the ligand for the ILT7 receptor.