missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD314 (NKG2D) Rat anti-Mouse, PE-eFluor 610, Clone: CX5, eBioscience™

Rat Monoclonal Antibody

$158.05 - $381.48

Specifications

Antigen CD314 (NKG2D)
Clone CX5
Host Species Rat
Gene Alias KLRK1
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501124842 Life Technologies
LS61588280
25μg Each for $158.05
501124843 Life Technologies
LS61588282
100μg Each for $381.48
Specifications

Description

The CX5 monoclonal antibody reacts with the mouse NKG2D, a lectin-like molecule expressed on both human and mouse NK cells. Mouse NKG2D binds to retinoic acid-inducible RAE-1 alpha, beta, gamma, delta, epsilon and the minor histocompatibility molecule H60 and has the ability to costimulate multiple NK activation receptors, through the DAP12/DAP10 adaptor molecules. NKG2D is expressed by all spleen and liver NK cells, NK1.1+ thymocytes, in vitro activated LAK cells, and a subset of splenic NKT cells.
Description & Specifications

Specifications

CD314 (NKG2D)
Rat
Flow Cytometry
PE-eFluor 610
Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Monoclonal
0.2mg/mL
CX5
KLRK1
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer