Please login to your online account to display your discounted pricing

CD314 (NKG2D) Rat anti-Mouse, PE-Cy7, Clone: CX5, eBioscience™

Rat Monoclonal Antibody

$245.05 - $419.05

Specifications

Antigen CD314 (NKG2D)
Clone CX5
Host Species Rat
Gene Alias KLRK1
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5015586

 
life technologies
LS25588281
50μg Each for $245.05

5015587

 
life technologies
LS25588282
100μg Each for $419.05
Description & Specifications

Specifications

Antigen CD314 (NKG2D)
Clone CX5
Host Species Rat
Gene Alias KLRK1
Species Reactivity Mouse
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE-Cy7
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The CX5 monoclonal antibody reacts with the mouse NKG2D, a lectin-like molecule expressed on both human and mouse NK cells. Mouse NKG2D binds to retinoic acid-inducible RAE-1 alpha, beta, gamma, delta, epsilon and the minor histocompatibility molecule H60 and has the ability to costimulate multiple NK activation receptors, through the DAP12/DAP10 adaptor molecules. NKG2D is expressed by all spleen and liver NK cells, NK1.1+ thymocytes, in vitro activated LAK cells, and a subset of splenic NKT cells.