Please login to your online account to display your discounted pricing

CD314 (NKG2D) Rat anti-Mouse, Clone: MI-6 (MI6), Functional Grade, eBioscience™

Rat Monoclonal Antibody, Functional Grade

$259.55 - $694.55

Specifications

Antigen CD314 (NKG2D)
Clone MI-6 (MI6)
Host Species Rat
Gene Alias KLRK1
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5014591

 
affymetrix
16588082
100μg Each for $259.55

5014592

 
affymetrix
16588085
500μg Each for $419.05

5014593

 
affymetrix
16588086
1mg Each for $694.55
Description & Specifications

Specifications

Antigen CD314 (NKG2D)
Clone MI-6 (MI6)
Host Species Rat
Gene Alias KLRK1
Species Reactivity Mouse
Applications Flow Cytometry
Applications Functional Assay
Regulatory Status RUO
Conjugate Unlabeled
Format Purified
Storage Requirements Store at 2-8°C.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, no sodium azide
Concentration 1mg/mL

The MI-6 monoclonal antibody reacts with the mouse NKG2D, a lectin-like molecule expressed on both human and mouse NK cells. Mouse NKG2D binds to retinoic acid-inducible RAE-1 alpha, beta, gamma, delta, epsilon and the minor histocompatibility molecule H60 and has the ability to costimulate multiple NK activation receptors, through the DAP12/DAP10 adaptor molecules. NKG2D is expressed by all spleen and liver NK cells, NK1.1(+) thymocytes, in vitro activated LAK cells, and a subset of splenic NKT cells.