Please login to your online account to display your discounted pricing

CD314 (NKG2D) Mouse anti-Human, PerCP-eFluor 710, Clone: 1D11, eBioscience™

Mouse Monoclonal Antibody

$172.55 - $442.25

Specifications

Antigen CD314 (NKG2D)
Clone 1D11
Host Species Mouse
Gene Alias KLRK1
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501123850

 
life technologies
LS46587841
25 tests Each for $172.55

501123851

 
life technologies
LS46587842
100 tests Each for $442.25
Description & Specifications

Specifications

Antigen CD314 (NKG2D)
Clone 1D11
Host Species Mouse
Gene Alias KLRK1
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PerCP-eFluor 710
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.125μg)/test

The 1D11 monoclonal antibody reacts with the human NKG2D, a 42kDa lectin-like molecule expressed by NK cells, gamma delta T cells, CD8+ T cells, and some CD4+ T cells. Human NKG2D forms complexes with DAP10, a membrane adaptor protein, and has the ability to costimulate multiple NK activation receptors. The counter-receptor for human NKG2D has been identified as MICA/MICB expressed on epithelial tumors from lung, breast, kidney, ovary, prostate and colon carcinoma. 5C6 and 1D11 block binding of soluble MICA to gamma delta TCR T cell clones and inhibit lysis by these cells. 5C6 and 1D11 induced NKG2D function of redirected lysis of FcReceptor bearing P815 cells.