missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD314 (NKG2D) Mouse anti-Human, APC, Clone: 1D11, eBioscience™

Mouse Monoclonal Antibody

$361.05 - $390.05

Specifications

Antigen CD314 (NKG2D)
Clone 1D11
Host Species Murine
Gene Alias KLRK1
Species Reactivity Human
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501123151 Life Technologies
LS17587842
100 Tests Each for $390.05
5015229 Life Technologies
LS17587881
50μg Each for $361.05
Specifications

Description

The 1D11 monoclonal antibody reacts with the human NKG2D, a 42kDa lectin-like molecule expressed by NK cells, gamma delta T cells, CD8+ T cells, and some CD4+ T cells. Human NKG2D forms complexes with DAP10, a membrane adaptor protein, and has the ability to costimulate multiple NK activation receptors. The counter-receptor for human NKG2D has been identified as MICA/MICB expressed on epithelial tumors from lung, breast, kidney, ovary, prostate and colon carcinoma. 5C6 and 1D11 block binding of soluble MICA to gamma delta TCR T cell clones and inhibit lysis by these cells. 5C6 and 1D11 induced NKG2D function of redirected lysis of FcReceptor bearing P815 cells.
Description & Specifications

Specifications

CD314 (NKG2D)
Murine
Human
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
1D11
KLRK1
Flow Cytometry
Allophycocyanin
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.2mg/mL; 5μL (0.5μg)/test