Please login to your online account to display your discounted pricing

CD31 (PECAM-1) Rat anti-Mouse, PE-Cy7, Clone: 390, eBioscience™

Rat Monoclonal Antibody

$158.05 - $294.35

Specifications

Antigen CD31 (PECAM-1)
Clone 390
Host Species Rat
Gene Alias Platelet-Endothelial Cell Adhesion Molecule 1
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5015497

 
life technologies
LS25031181
50μg Each for $158.05

5015498

 
life technologies
LS25031182
100μg Each for $294.35
Description & Specifications

Specifications

Antigen CD31 (PECAM-1)
Clone 390
Host Species Rat
Gene Alias Platelet-Endothelial Cell Adhesion Molecule 1
Species Reactivity Mouse
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE-Cy7
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The 390 monoclonal antibody reacts with mouse CD31, also known as platelet-endothelial cell adhesion molecule-1 (PECAM-1) and gpIIa. This 130-140kDa surface protein is expressed by endothelial cells and at low levels on all leukocytes and platelets. It has been reported that CD38 binds to CD31. Homotypic interaction of CD31 is important in adhesion, cell-cell and cell-matrix interaction, and signal transduction.