missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD31 (PECAM-1) Rat anti-Mouse, FITC, Clone: 390, eBioscience™

Rat Monoclonal Antibody

$130.50 - $397.30

Specifications

Antigen CD31 (PECAM-1)
Clone 390
Host Species Rat
Gene Alias Platelet-Endothelial Cell Adhesion Molecule 1
Species Reactivity Mouse
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
509517 Life Technologies
LS11031181
50μg Each for $130.50
501128997 Life Technologies
LS11031182
100μg Each for $205.90
501129390 Life Technologies
LS11031185
500μg Each for $397.30
Specifications

Description

The 390 monoclonal antibody reacts with mouse CD31, also known as platelet-endothelial cell adhesion molecule-1 (PECAM-1) and gpIIa. This 130-140kDa surface protein is expressed by endothelial cells and at low levels on all leukocytes and platelets. It has been reported that CD38 binds to CD31. Homotypic interaction of CD31 is important in adhesion, cell-cell and cell-matrix interaction, and signal transduction.
Description & Specifications

Specifications

CD31 (PECAM-1)
Rat
Mouse
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
390
Platelet-Endothelial Cell Adhesion Molecule 1
Flow Cytometry,Immunohistochemistry (Frozen)
FITC
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.5mg/mL