missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD31 (PECAM-1), PE-Cyanine7, clone: TLD-3A12, eBioscience™

Mouse Monoclonal Antibody

Manufacturer:  Life Technologies LS25031080

 View more versions of this product

Catalog No. 501129263



Description

Description

CD31 (platelet endothelial cell adhesion molecule-1, PECAM-1) is an inhibitory coreceptor involved in regulation of T cell and B cell signaling by a dual immunoreceptor tyrosine-based inhibitory motif (ITIM) that upon associated kinases-mediated phosphorylation provide docking sites for protein-tyrosine phosphatases. CD31 is expressed ubiquitously within the vascular compartment and is located mainly at junctions between adjacent cells. N-terminal Ig-like domain of CD31 is responsible for its homophilic binding, which plays an important role in cell-cell interactions. CD31 is a multifunctional molecule with diverse roles in modulation of integrin-mediated cell adhesion, transendothelial migration, angiogenesis, apoptosis, negative regulation of immunoreceptor signaling, autoimmunity, macrophage phagocytosis, IgE-mediated anaphylaxis and thrombosis. It is one of key regulatory molecules in vascular system.

Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically. Excitation: 488-561 nm; Emission: 775 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered. CD31 (platelet endothelial cell adhesion molecule-1, PECAM-1) is an inhibitory coreceptor involved in regulation of T cell and B cell signaling by a dual immunoreceptor tyrosine-based inhibitory motif (ITIM) that upon associated kinases-mediated phosphorylation provide docking sites for protein-tyrosine phosphatases. CD31 is expressed ubiquitously within the vascular compartment and is located mainly at junctions between adjacent cells. N-terminal Ig-like domain of CD31 is responsible for its homophilic binding, which plays an important role in cell-cell interactions. CD31 is a multifunctional molecule with diverse roles in modulation of integrin-mediated cell adhesion, transendothelial migration, angiogenesis, apoptosis, negative regulation of immunoreceptor signaling, autoimmunity, macrophage phagocytosis, IgE-mediated anaphylaxis and thrombosis. It is one of key regulatory molecules in vascular system.
Specifications

Specifications

CD31 (PECAM-1)
Monoclonal
0.2 mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
Q3SWT0
CD31 antigen, CD31/EndoCAM, GPIIA′, PECA1, PECAM-1, CD31/EndoCAM, adhesion molecule, endoCAM, platelet endothelial cell adhesion molecule
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Rat
Flow Cytometry
TLD-3A12
PE-Cyanine7
Pecam1
Liquid
Pecam1
IgG1, kappa
25 μg
4° C, store in dark, DO NOT FREEZE!
Primary
29583
Documents

For Research Use Only.