Please login to your online account to display your discounted pricing

CD31 (PECAM-1) Mouse anti-Rat, eFluor 660, Clone: TLD-3A12, eBioscience™

Mouse Monoclonal Antibody

$129.05 - $317.55

Specifications

Antigen CD31 (PECAM-1)
Clone TLD-3A12
Host Species Mouse
Gene Alias Platelet endothelial cell adhesion molecule, platelet membrane GPIIa
Species Reactivity Rat
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501124264

 
life technologies
LS50031080
25μg Each for $129.05

501124265

 
life technologies
LS50031082
100μg Each for $317.55
Description & Specifications

Specifications

Antigen CD31 (PECAM-1)
Clone TLD-3A12
Host Species Mouse
Gene Alias Platelet endothelial cell adhesion molecule, platelet membrane GPIIa
Species Reactivity Rat
Applications Flow Cytometry
Regulatory Status RUO
Conjugate eFluor 660
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The TLD-3A12 monoclonal antibody reacts with rat CD31, also known as platelet-endothelial cell adhesion molecule-1 (PECAM-1) and gpIIa. This 130kDa surface protein is expressed highly by endothelial cells but at low levels on all leukocytes and platelets. Studies have demonstrated that CD38 binds to CD31. Homotypic interaction of CD31 is important in adhesion, cell-cell and cell-matrix interactions, and signal transduction.