Please login to your online account to display your discounted pricing

CD31 (PECAM-1) Mouse anti-Human, PE-Cy7, Clone: WM-59 (WM59), eBioscience™

Mouse Monoclonal Antibody

$187.05 - $426.30

Specifications

Antigen CD31 (PECAM-1)
Clone WM-59 (WM59)
Host Species Mouse
Gene Alias Platelet-Endothelial Cell Adhesion Molecule 1
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

50245725

 
life technologies
LS25031941
25 tests Each for $187.05

50245726

 
life technologies
LS25031942
100 tests Each for $426.30
Description & Specifications

Specifications

Antigen CD31 (PECAM-1)
Clone WM-59 (WM59)
Host Species Mouse
Gene Alias Platelet-Endothelial Cell Adhesion Molecule 1
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE-Cy7
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.06μg)/test

The WM59 monoclonal antibody reacts with human CD31, also known as platelet-endothelial cell adhesion molecule-1 (PECAM-1) and gpIIa. This 130-140kDa surface protein is expressed by endothelial cells and at low levels on leukocytes and platelets. It has been reported that CD38 binds to CD31. Homotypic interaction of CD31 is important in adhesion, cell-cell and cell-matrix interaction, and signal transduction.