missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD31 (PECAM-1) Mouse anti-Human, PE, Clone: WM-59 (WM59), eBioscience™

Mouse Monoclonal Antibody

$130.68 - $302.28

Specifications

Antigen CD31 (PECAM-1)
Clone WM-59 (WM59)
Host Species Murine
Gene Alias Platelet-Endothelial Cell Adhesion Molecule 1
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5010323 Life Technologies
LS12031942
100 Tests Each for $302.28
5010322 Life Technologies
LS12031941
25 Tests Each for $130.68
Specifications

Description

The WM59 monoclonal antibody reacts with human CD31, also known as platelet-endothelial cell adhesion molecule-1 (PECAM-1) and gpIIa. This 130-140kDa surface protein is expressed by endothelial cells and at low levels on leukocytes and platelets. It has been reported that CD38 binds to CD31. Homotypic interaction of CD31 is important in adhesion, cell-cell and cell-matrix interaction, and signal transduction.
Description & Specifications

Specifications

CD31 (PECAM-1)
Murine
Flow Cytometry
PE
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
5μL (0.125μg)/test
WM-59 (WM59)
Platelet-Endothelial Cell Adhesion Molecule 1
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer