missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD31 (PECAM-1) Mouse anti-Human, FITC, Clone: WM-59 (WM59), eBioscience™

Mouse Monoclonal Antibody

$104.28 - $236.28

Specifications

Antigen CD31 (PECAM-1)
Clone WM-59 (WM59)
Host Species Murine
Gene Alias Platelet-Endothelial Cell Adhesion Molecule 1
Species Reactivity Human
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
509520 Life Technologies
LS11031941
25 Tests Each for $104.28
509521 Life Technologies
LS11031942
100 Tests Each for $236.28
Specifications

Description

The WM59 monoclonal antibody reacts with human CD31, also known as platelet-endothelial cell adhesion molecule-1 (PECAM-1) and gpIIa. This 130-140kDa surface protein is expressed by endothelial cells and at low levels on leukocytes and platelets. It has been reported that CD38 binds to CD31. Homotypic interaction of CD31 is important in adhesion, cell-cell and cell-matrix interaction, and signal transduction.
Description & Specifications

Specifications

CD31 (PECAM-1)
Murine
Human
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
WM-59 (WM59)
Platelet-Endothelial Cell Adhesion Molecule 1
Flow Cytometry
FITC
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
5μL (0.5μg)/test