Please login to your online account to display your discounted pricing

CD31 (PECAM-1) Mouse anti-Human, Clone: WM-59 (WM59), eBioscience™

Mouse Monoclonal Antibody

$71.05 - $158.05

Specifications

Antigen CD31 (PECAM-1)
Clone WM-59 (WM59)
Host Species Mouse
Gene Alias Platelet-Endothelial Cell Adhesion Molecule 1
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5012523

 
life technologies
LS14031980
25μg Each for $71.05

5012524

 
life technologies
LS14031982
100μg Each for $158.05
Description & Specifications

Specifications

Antigen CD31 (PECAM-1)
Clone WM-59 (WM59)
Host Species Mouse
Gene Alias Platelet-Endothelial Cell Adhesion Molecule 1
Species Reactivity Human
Applications Flow Cytometry
Applications Immunohistochemistry (Frozen)
Regulatory Status RUO
Conjugate Unlabeled
Format Purified
Storage Requirements Store at 2-8°C.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

The WM59 monoclonal antibody reacts with human CD31, also known as platelet-endothelial cell adhesion molecule-1 (PECAM-1) and gpIIa. This 130-140kDa surface protein is expressed by endothelial cells and at low levels on leukocytes and platelets. It has been reported that CD38 binds to CD31. Homotypic interaction of CD31 is important in adhesion, cell-cell and cell-matrix interaction, and signal transduction.