missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ CD31/PECAM-1 (Endothelial Cell Marker), Rabbit Polyclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry procedures with Thermo Scientific™ CD31/PECAM-1 (Endothelial Cell Marker), Rabbit Polyclonal Antibody.

$283.22 - $1,172.27

Specifications

Antigen CD31/PECAM-1 (Endothelial Cell Marker)
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unlabeled
Cross Reactivity Human
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
RB10333P Thermo Scientific™ Lab Vision™
RB10333P
1mL Each for $1,172.27
RB10333P0 Thermo Scientific™ Lab Vision™
RB10333P0
100μL Each for $283.22
RB10333P1 Thermo Scientific™ Lab Vision™
RB10333P1
500μL Each for $704.12
RB10333R7 Thermo Scientific™ Lab Vision™
RB10333R7
7mL Each for $589.88
Specifications

Description

Ensure accurate, reproducible results in immunohistochemistry procedures with Thermo Scientific™ CD31/PECAM-1 (Endothelial Cell Marker), Rabbit Polyclonal Antibody.

CD31 is a glycoprotein expressed on endothelial cells and in platelets. It is known to be involved in cell signaling and cell adhesion. Antibody to CD31 is of value in the study of benign and malignant vascular tumors. Staining for CD31 has also been used to measure angiogenesis, which reportedly predicts tumor recurrence.

Host Species: Rabbit

Species Reactivity: Human. Others not tested.

Epitope: C-terminal

Immunogen: Synthetic peptide corresponding to C-terminus of mouse CD31 protein

Molecular Weight: 100 and 130 kDa

Positive Control: Tonsil, Placenta

Cellular Localization: Endothelial cells in blood vessel of tonsil

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO

Description & Specifications

Specifications

CD31/PECAM-1 (Endothelial Cell Marker)
Polyclonal
Human
RUO
Primary
Tonsil
Immunohistochemistry (Paraffin)
Unlabeled
Rabbit
Cancer and Tumor Biology
Synthetic peptide corresponding to C-terminus of mouse CD31 protein