missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD309 (FLK1) Rat anti-Mouse, Alexa Fluor 700, Clone: Avas12a1, eBioscience™

Rat Monoclonal Antibody

$223.08 - $394.68

Specifications

Antigen CD309 (FLK1)
Clone Avas12a1
Host Species Rat
Gene Alias Flk-1, VEGF-R2, VEGFR2, Ly-73, Ly73, KDR
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5016921 Life Technologies
LS56582180
25μg Each for $223.08
5016922 Life Technologies
LS56582181
50μg Each for $394.68
Specifications

Description

The Avas12a1 monoclonal antibody reacts with mouse Flk-1, also known as vascular endothelial growth factor receptor 2 (VEGFR2). Flk-1 is a receptor tyrosine kinase involved in vascular endothelial tissue development and is expressed on endothelial cells during embryonic stages and some endothelial tissues in the adult.
Description & Specifications

Specifications

CD309 (FLK1)
Rat
Flow Cytometry
Alexa Fluor 700
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.2mg/mL
Avas12a1
Flk-1, VEGF-R2, VEGFR2, Ly-73, Ly73, KDR
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer