Please login to your online account to display your discounted pricing

CD304 (Neuropilin-1) Rat anti-Mouse, eFluor 450, Clone: 3DS304M, eBioscience™

Rat Monoclonal Antibody

$216.05 - $520.55

Specifications

Antigen CD304 (Neuropilin-1)
Clone 3DS304M
Host Species Rat
Gene Alias Nrp-1, NP-1, BDCA-4
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501124150

 
affymetrix
48304180
25μg Each for $216.05

501128640

 
affymetrix
48304182
100μg Each for $520.55
Description & Specifications

Specifications

Antigen CD304 (Neuropilin-1)
Clone 3DS304M
Host Species Rat
Gene Alias Nrp-1, NP-1, BDCA-4
Species Reactivity Mouse
Applications Flow Cytometry
Regulatory Status RUO
Conjugate eFluor 450
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The monoclonal antibody 3DS304M recognizes mouse CD304 (Neuropilin-1), a type 1 transmembrane glycoprotein present on the surface of multiple cell types, including: regulatory T cells (Treg), NKT cells, DC, certain types of stem cells, neurons, endothelial cells, and some neoplastic cells. Neuropilin-1, in complex with plexins, serves as a co-receptor for type 3 semaphorins in growing neurons. It is also involved in the process of angiogenesis being a part of a functional receptor for VEGF in endothelial cells. In mice, Neuropilin-1 is expressed on thymus-derived natural Treg but not peripherally induced Treg. In addition, transient expression on recently activated non-regulatory T cells has been observed. Neuropilin-1 has been used as a marker of recent thymic emigrants in the mouse iNKT cell population. It has been shown that Neuropilin-1 forms a complex with TGF beta receptors, activating the latent form of TGF beta (LAP-TGF beta 1) and augmenting canonical Smad2/3 signaling.

This 3DS304M antibody will recognize formaldehyde-fixed as well as methanol-treated epitopes.