Please login to your online account to display your discounted pricing

CD304 (Neuropilin-1) Rat anti-Mouse, Clone: XML1Y1, eBioscience™

Rat Monoclonal Antibody

$160.95 - $339.30

Specifications

Antigen CD304 (Neuropilin-1)
Clone XML1Y1
Host Species Rat
Gene Alias Nrp-1, NP-1, BDCA-4
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501122670

 
life technologies
LS14304280
25μg Each for $160.95

501122671

 
life technologies
LS14304282
100μg Each for $339.30
Description & Specifications

Specifications

Antigen CD304 (Neuropilin-1)
Clone XML1Y1
Host Species Rat
Gene Alias Nrp-1, NP-1, BDCA-4
Species Reactivity Mouse
Applications Immunocytochemistry
Applications Immunohistochemistry (Frozen)
Regulatory Status RUO
Conjugate Unlabeled
Format Purified
Storage Requirements Store at 2-8°C.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

The monoclonal antibody XML1Y1 recognizes CD304 (Neuropilin-1), a type 1 transmembrane glycoprotein present on the surface of multiple cell types, including: regulatory T cells (Tregs), NKT cells, DC, certain types of stem cells, neurons, endothelial cells, and some neoplastic cells. Neuropilin-1, in complex with plexins, serves as a co-receptor for type 3 semaphorins in growing neurons. It is also involved in the process of angiogenesis being a part of a functional receptor for VEGF in endothelial cells. In mouse Neuropilin-1 is expressed on thymus-derived natural Tregs but not peripherally induced Treg cells. In addition, transient expression on recently activated non-regulatory T cells has been observed. Neuropilin-1 has been used as a marker of recent thymic emigrants in the mouse iNKT cell population. It has been shown that Neuropilin-1 forms a complex with TGF beta receptors, activating TGF beta latent form (LAP-TGF beta 1) and augmenting canonic Smad2/3 signaling.

This XML1Y1 antibody recognizes neuropilin-1 isoforms 1 and 2.