missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD304 Mouse anti-Human, PE-eFluor 610, Clone: TNKUSOHA, eBioscience™

Mouse Monoclonal Antibody

$209.88 - $500.28

Specifications

Antigen CD304
Clone TNKUSOHA
Host Species Murine
Gene Alias BDCA-4, Neuropilin-1
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501124803 Life Technologies
LS61304942
100 Tests Each for $500.28
501124802 Life Technologies
LS61304941
25 Tests Each for $209.88
Specifications

Description

The TNKUSOHA monoclonal antibody recognizes human CD304, which is also known as BDCA-4 and Neuropilin-1. Like CD303, the CD304 antigen is expressed on human plasmacytoid dendritic cells in normal blood and bone marrow. CD304 can also be detected on T follicular helper cells in inflamed tonsils, as well as on some B-cell acute lymphoblastic leukemias. In addition, CD304 is expressed on numerous non-hematopoietic cell types, including neurons, endothelial, and tumor cells. Plasmacytoid dendritic cells play a key role in the innate immune response against viral pathogens and tumors.
Description & Specifications

Specifications

CD304
Murine
Flow Cytometry
PE-eFluor 610
Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Monoclonal
5μL (0.25μg)/test
TNKUSOHA
BDCA-4, Neuropilin-1
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer